Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700032166.mm.1 |
NM_198831.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032166.mm.1 |
NM_198831.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032166.mm.1 |
NR_003559.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032166.mm.1 |
NR_003559.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032166.mm.1 |
TC0700003822.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700032165.mm.1 |
NM_198831.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032165.mm.1 |
NM_198831.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032165.mm.1 |
NR_003559.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032165.mm.1 |
NR_003559.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700032165.mm.1 |
TC0700003822.mm.1 |
MTA TC |