Assay Details
Gene Symbol: Cox6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053071.2 | NP_444301.1 | 2 - 3 | 256 | 96 | |
GenBank mRNA | AK012602.1 | - | 2 - 3 | 173 | 96 | |
AK013459.1 | - | 2 - 3 | 172 | 96 | ||
BC024666.1 | - | 2 - 3 | 187 | 4195595 | 96 | |
BC094413.1 | - | 2 - 3 | 159 | 1397127 | 96 | |
BY100624.1 | - | 2 - 3 | 258 | 96 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30135996 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30135994 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30135995 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30135997 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30135998 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30136001 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30136000 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P343323 | AK012602.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P343323 | AK012602.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 822464 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 945268 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 479664 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491711 | AJ131357.2 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491711 | AK019478.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491711 | AK050615.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491711 | AK153975.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491711 | BC132166.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491711 | BC132466.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491711 | NM_009913.5 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 160131 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P144778 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P343323 | AK012602.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P343323 | AK012602.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P343323 | AK012602.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P343323 | AK012602.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30135991 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30135998 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30136007 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30135985 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30136000 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30135995 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30136006 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30135983 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30135987 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30136003 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30136011 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30135997 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30135981 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30135989 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30136009 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30136015 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30135993 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P144778 | - | - |
Whole Mouse Genome G4122A | A_51_P343323 | AK012602.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P343323 | AK012602.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132417 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P144778 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11920136 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11920145 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11920139 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11920117 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11920144 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11920133 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11920124 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30136001 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30136000 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30135995 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30135997 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30135994 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30135996 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30135998 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135926 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136014 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135947 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135991 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135919 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135892 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135906 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135989 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135952 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135936 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135983 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136003 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135908 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135901 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135921 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135922 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135895 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135993 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135925 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135930 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135985 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135975 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135965 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135887 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135981 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135937 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135939 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135903 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135899 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135915 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135977 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135960 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136013 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135897 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135962 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135944 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135912 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135896 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135911 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135918 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135954 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135894 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135997 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135949 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135998 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135927 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135932 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135924 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136000 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136025 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135888 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135891 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135941 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30136006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135957 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135946 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135955 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30135914 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135908 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135924 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135927 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135944 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12327137 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135913 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135935 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12269141 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135975 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135985 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135894 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135995 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11920119 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12443264 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12443267 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135904 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135925 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11896479 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11920132 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30136019 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30136015 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30136003 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30136007 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135888 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135891 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135934 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135901 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135919 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12379478 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135903 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135922 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12393722 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135941 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135972 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135993 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135991 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135915 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135949 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135899 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135960 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135921 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135937 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135989 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11920139 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12269146 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135981 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12443266 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135983 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30136009 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135906 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135885 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135962 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30136013 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135911 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135896 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135887 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30136018 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12443290 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135916 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135955 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135957 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135892 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135918 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135968 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30136014 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30136017 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135936 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135987 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135965 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135970 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11920144 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135977 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135947 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135973 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30135980 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30136016 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10428121 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10400473 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10428120 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10428123 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10400472 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10400471 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10428122 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1434491_PM_a_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1415970_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10400470 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10428119 | - | - |
Mouse Genome 430 2.0 Array | 1434491_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1434491_a_at | - | - |
Mouse Genome 430 2.0 Array | 1415970_at | - | - |
Mouse Genome 430 2.0 Array | 1415970_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12864_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12864_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1500010498.mm.1 | NM_053071.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500010498.mm.1 | NM_053071.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500010498.mm.1 | TC1500001365.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500010500.mm.1 | NM_053071.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500010500.mm.1 | NM_053071.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500010500.mm.1 | TC1500001365.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500010501.mm.1 | NM_053071.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500010501.mm.1 | NM_053071.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500010501.mm.1 | TC1500001365.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500010499.mm.1 | NM_053071.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500010499.mm.1 | NM_053071.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500010499.mm.1 | TC1500001365.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500010510.mm.1 | NM_053071.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500010510.mm.1 | NM_053071.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500010510.mm.1 | TC1500001365.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500010511.mm.1 | NM_053071.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500010511.mm.1 | NM_053071.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500010511.mm.1 | TC1500001365.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500010503.mm.1 | NM_053071.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500010503.mm.1 | NM_053071.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500010503.mm.1 | TC1500001365.mm.1 | MTA TC |
Mouse Expression Set 430 | 1434491_PM_a_at | - | - |
Mouse Expression Set 430 | 1434491_a_at | - | - |
Mouse Expression Set 430 | 1415970_at | - | - |
Mouse Expression Set 430 | 1415970_PM_at | - | - |
Murine Genome U74v2 Set | 100551_PM_r_at | - | - |
Murine Genome U74v2 Set | 100550_PM_f_at | - | - |
Murine Genome U74v2 Set | 100550_f_at | - | - |
Murine Genome U74v2 Set | 100551_r_at | - | - |
Murine Genome U74v2 Set | 162319_PM_i_at | - | - |
Murine Genome U74v2 Set | 162319_i_at | - | - |
Mouse Genome 430A 2.0 Array | 1434491_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1415970_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1434491_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1415970_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1415970_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1434491_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1415970_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1434491_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 1400020 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001400020 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2589039 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2589039 | - | - |
mouse-6 v1.1 (Illumina) | scl012864.3_58-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1400020 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl012864.3_58-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1400020 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2589039 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2589039 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl012864.3_58-S | - | - |
