Assay Details
Gene Symbol: Kifc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195298.1 | NP_001182227.1 | 9 - 10 | 2041 | 208 | |
GenBank mRNA | AF221102.1 | - | 9 - 10 | 2024 | 208 | |
AI326799.1 | - | 2 - 3 | 208 | |||
AK133900.1 | - | 9 - 10 | 2047 | 208 | ||
BC003753.1 | - | 9 - 10 | 1952 | 3495308 | 208 | |
BC057162.1 | - | 9 - 10 | 1977 | 5350661 | 208 | |
BC100328.1 | - | 9 - 10 | 1996 | 30841951 | 208 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
