Assay Details
| Gene Symbol: Vmn1r44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053227.2 | NP_444457.2 | 1 | 561 | 155 | |
| GenBank mRNA | BC107184.1 | - | 1 | 561 | 155 | |
| BC107185.1 | - | 1 | 561 | 155 | ||
| BC151088.1 | - | 1 | 658 | 9088001 | 155 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2014839 | BC107184.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2014839 | BC107185.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2014839 | BC151088.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2014839 | NM_053227.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2014839 | BC107184.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2014839 | BC107185.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2014839 | BC151088.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2014839 | NM_053227.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P437906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630267 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630236 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24630245 | - | - |
| Whole Mouse Genome G4122A | A_51_P437906 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05488044 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05082741 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05645835 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05488039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24630226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24630245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05876244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05645835 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05876249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24630266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24630223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24630283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24630292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05749680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24630256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24630284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24630236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05876253 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24630275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05048856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24630288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24630294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24630259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24630229 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24630231 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05645838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05749688 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05312744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24630220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24630230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24630279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24630228 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05082741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24630233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05660761 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24630244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24630257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24630270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05876258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24630263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05360793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05048851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24630281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24630222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24630269 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10539959 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539958 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422377_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10539957 | - | - |
| Mouse Genome 430 2.0 Array | 1422377_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422377_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600008246.mm.1 | TC0600001055.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422377_at | - | - |
| Mouse Expression Set 430 | 1422377_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422377_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422377_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422377_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422377_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 100380280 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 100380280 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1050280 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001050280 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2492216 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2492216 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1221926 | - | - |
| mouse-6 v1.1 (Illumina) | scl0113854.1_103-S | - | - |
| mouse-6 v1.1 (Illumina) | GI_16716536-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1050280 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100380280 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0113854.1_103-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100380280 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1050280 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2492216 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2492216 | - | - |

Vmn1r44
Genomic Map