Assay Details
| Gene Symbol: Vmn1r205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134217.1 | NP_598978.1 | 1 | 903 | 186 | |
| GenBank mRNA | BC132016.1 | - | 1 | 973 | 40130293 | 186 |
| BC138103.1 | - | 1 | 973 | 8861122 | 186 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P227627 | BC138103.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P227627 | NM_134217.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P227627 | BC132016.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P227627 | BC138103.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P227627 | NM_134217.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P227627 | BC132016.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P227627 | BC138103.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P227627 | NM_134217.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P227627 | BC132016.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P227627 | BC138103.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P227627 | NM_134217.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P227627 | BC132016.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P227627 | BC138103.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P227627 | NM_134217.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P227627 | BC132016.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P227627 | BC138103.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P227627 | NM_134217.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P227627 | BC132016.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28878188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28878209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28878217 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28878219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28878185 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28878197 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28878201 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28878212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28878198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28878204 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28878210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28878225 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28878215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28878220 | - | - |
| Whole Mouse Genome G4122A | A_51_P227627 | BC138103.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P227627 | NM_134217.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P227627 | BC132016.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P227627 | BC138103.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P227627 | NM_134217.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P227627 | BC132016.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120984 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105894 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10499093 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10499100 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10499115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28878220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28878210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28878197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28878207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28878219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28878188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28878196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28878212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28878214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28878223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28878217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28878201 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10499084 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28878214 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28878197 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28878219 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28878178 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28878201 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10499112 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28878188 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11121792 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28878207 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28878204 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28878225 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28878185 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28878182 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28878200 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28878187 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28878180 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28878212 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10895176 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28878223 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10499100 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28878217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10663884 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28878210 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11105315 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10408149 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408145 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408141 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10408140 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300009960.mm.1 | NM_134217.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300009960.mm.1 | NM_134217.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300009960.mm.1 | TC1300001593.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300009961.mm.1 | TC1300001593.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300009959.mm.1 | TC1300001593.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 302207 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5860348 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5860348 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 100730136 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005860348 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2521809 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2521809 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1215539 | - | - |
| mouse-6 v1.1 (Illumina) | GI_21717750-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0171251.1_169-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5860348 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100730136 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0171251.1_169-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100730136 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5860348 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2906929 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2906929 | - | - |

Vmn1r205
Genomic Map