Assay Details
| Gene Symbol: Vmn1r193 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134225.1 | NP_598986.1 | 1 | 745 | 166 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P263719 | NM_134225.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P263719 | NM_134225.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P263719 | NM_134225.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P263719 | NM_134225.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P263719 | NM_134225.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P263719 | NM_134225.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28876859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28876864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28876836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28876860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28876834 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28876872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28876876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28876886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28876863 | - | - |
| Whole Mouse Genome G4122A | A_51_P263719 | NM_134225.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P263719 | NM_134225.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127348 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10992998 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10865341 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10486483 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10486485 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10486489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28876863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28876864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28876821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28876819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28876860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28876830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28876832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28876828 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28876834 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28876819 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28876848 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28876860 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28876821 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28876875 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28876876 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10651849 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10674183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28876864 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10902888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28876886 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28876866 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28876832 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28876859 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28876836 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10902879 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28876872 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10486486 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28876830 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10992998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28876882 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28876887 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10408135 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404005 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404023 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10404022 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10408134 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10404004 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300009953.mm.1 | NM_134225.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300009953.mm.1 | NM_134225.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300009953.mm.1 | TC1300001589.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 590591 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3440088 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003440088 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2516844 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2516844 | - | - |
| mouse-6 v1.1 (Illumina) | scl11522.1.1_89-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3440088 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl11522.1.1_89-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3440088 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2516844 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2516844 | - | - |

Vmn1r193
Genomic Map