Assay Details
| Gene Symbol: Oas1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145211.2 | NP_660212.2 | 2 - 3 | 519 | 159 | |
| GenBank mRNA | AK149613.1 | - | 2 - 3 | 480 | 159 | |
| BC013715.1 | - | 2 - 3 | 501 | 3257027 | 159 | |
| BC057878.1 | - | 2 - 3 | 478 | 5348858 | 159 | |
| M33863.1 | - | 505 | 159 | |||
| X04958.1 | - | 2 - 3 | 505 | 159 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1998942 | BC013715.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1998942 | BC057878.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1998943 | M33863.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1998943 | X04958.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1998943 | NM_145211.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1998943 | AK149613.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1998943 | BC013715.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 362554 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 533666 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 765221 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 76452 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1998942 | BC013715.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1998942 | BC057878.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1998943 | M33863.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1998943 | X04958.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1998943 | NM_145211.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1998943 | AK149613.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1998943 | BC013715.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041492 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041527 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041525 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041497 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041504 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041534 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05720379 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01204643 | - | - |
| Whole Mouse Genome G4122A | A_52_P337357 | BC013715.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P337357 | BC057878.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01204643 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04721673 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04139022 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04139023 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04362878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04721682 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04262110 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041505 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041468 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041504 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041492 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041473 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04721680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04262111 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041496 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24041499 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04312888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04839096 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10533260 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533259 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533258 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533249 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533250 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533251 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533248 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533255 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533253 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533254 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533257 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533247 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424775_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10533246 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10533256 | - | - |
| Mouse Genome 430 2.0 Array | 1424775_at | - | - |
| Mouse Genome 430 2.0 Array | 1424775_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500028273.mm.1 | TC0500003181.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500028273.mm.1 | NM_145211.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028273.mm.1 | NM_145211.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028271.mm.1 | TC0500003181.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500028271.mm.1 | NM_145211.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028271.mm.1 | NM_145211.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028277.mm.1 | TC0500003181.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500028277.mm.1 | NM_145211.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028277.mm.1 | NM_145211.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028276.mm.1 | TC0500003181.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500028276.mm.1 | NM_145211.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028276.mm.1 | NM_145211.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028270.mm.1 | TC0500003181.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500028270.mm.1 | NM_145211.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028270.mm.1 | NM_145211.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028272.mm.1 | TC0500003181.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500028272.mm.1 | NM_145211.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028272.mm.1 | NM_145211.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028274.mm.1 | TC0500003181.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500028274.mm.1 | NM_145211.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028274.mm.1 | NM_145211.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028275.mm.1 | TC0500003181.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500028275.mm.1 | NM_145211.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028275.mm.1 | NM_145211.2 | Refseq |
| Affymetrix HT Mouse Genome 430A Array | 1424775_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424775_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 839637 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4760048 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004760048 | - | - |
| mouse-6 v1.1 (Illumina) | scl00246730.1_27-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4760048 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00246730.1_27-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4760048 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2924839 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1255890 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2924839 | - | - |

Oas1a
Genomic Map