Assay Details
| Gene Symbol: Olfr498 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146307.2 | NP_666419.2 | 1 | 253 | 157 | |
 Back To Top
 Back To TopMore Information
Additional Information:
				    	 
					    	
					        	
					            	Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
					        	
					    	            
				    	
			    	
			    
			| Set Membership: |  Ambion siRNA available  Amplicon length greater than or equal to 101  G-protein coupled receptor  Detect Genomic DNA  Recommended Assay  Mus musculus  Both primers and probe map within a single exon | 
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
 Back To Top
 Back To Top
 Olfr498
Olfr498 Genomic Map
Genomic Map Back To Top
 Back To Top