Assay Details
Gene Symbol: Olfr434 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146369.1 | NP_666481.1 | 1 | 434 | 188 | |
GenBank mRNA | BC120611.1 | - | 1 | 466 | 40129534 | 188 |
BC120613.1 | - | 1 | 466 | 40129536 | 188 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P286436 | NM_146369.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P286436 | BC120611.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P286436 | BC120613.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P286436 | NM_146369.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P286436 | BC120611.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P286436 | BC120613.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150426 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P286436 | NM_146369.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P286436 | BC120611.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P286436 | BC120613.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P286436 | NM_146369.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P286436 | BC120611.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P286436 | BC120613.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P286436 | NM_146369.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P286436 | BC120611.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P286436 | BC120613.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P286436 | NM_146369.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P286436 | BC120611.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P286436 | BC120613.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24402123 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24402112 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24402105 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24402128 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24402110 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24402106 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24402098 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24402095 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24402121 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24402081 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24402108 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24402122 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24402131 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24402129 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24402109 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24402097 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24402127 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24402092 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24402125 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24402075 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24402104 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24402099 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24402133 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24402102 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24402089 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24402100 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150426 | - | - |
Whole Mouse Genome G4122A | A_51_P286436 | NM_146369.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P286436 | BC120611.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P286436 | BC120613.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P286436 | NM_146369.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P286436 | BC120611.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P286436 | BC120613.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119259 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150426 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05805292 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05805296 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05805277 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05805271 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05805319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402092 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402129 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402123 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402102 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402074 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402104 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402131 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402057 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402060 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402105 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402095 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402127 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402055 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402097 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402145 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402066 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402133 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402128 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402140 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402098 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402064 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402069 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24402125 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402060 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402145 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402069 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402055 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402062 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05872601 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402074 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402112 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402108 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05805276 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402143 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402054 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402066 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402106 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402063 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05805317 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402102 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402140 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402129 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402105 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402075 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402128 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402137 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402110 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402121 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402104 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05805306 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402073 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402125 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402123 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402092 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05805291 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05604629 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402116 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402089 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402057 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402097 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402127 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402100 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402099 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402133 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402131 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402098 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05805302 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402065 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24402058 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10537830 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10537829 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10537828 | - | - |
Affymetrix Mouse Clariom S Array | PSR0600003778.mm.1 | TC0600000485.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 699512 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 4730014 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4730014 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004730014 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2701679 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2701679 | - | - |
mouse-6 v1.1 (Illumina) | scl30116.1.1_17-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4730014 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl30116.1.1_17-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4730014 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2701679 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2701679 | - | - |
