Assay Details
| Gene Symbol: Olfr291 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146415.2 | NP_666527.1 | 3 | 753 | 176 | |
| GenBank mRNA | BC100693.1 | - | 1 | 547 | 176 | |
| BC100694.1 | - | 1 | 546 | 176 | ||
| BC100695.1 | - | 1 | 547 | 176 | ||
| BC114974.1 | - | 1 | 547 | 40043317 | 176 | |
| BC115740.1 | - | 1 | 547 | 176 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2137531 | BC115740.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2137531 | BC100695.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2137531 | BC114974.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2137531 | NM_146415.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2137531 | BC100693.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2137531 | BC115740.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2137531 | BC100695.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2137531 | BC114974.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2137531 | NM_146415.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2137531 | BC100693.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P485531 | BC115740.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P485531 | BC100695.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P485531 | BC114974.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P485531 | NM_146415.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P485531 | BC100693.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P485537 | BC115740.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P485537 | NM_146416.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P485537 | BC100695.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P485537 | BC114974.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P485537 | BC127990.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25290628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25290621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25290625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25290631 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25290615 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25290629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25290611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25290612 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25290623 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25290617 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25290614 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25290640 | - | - |
| Whole Mouse Genome G4122A | A_51_P485531 | BC115740.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P485531 | BC100695.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P485531 | BC114974.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P485531 | NM_146415.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P485531 | BC100693.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P485537 | BC115740.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P485537 | NM_146416.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P485537 | BC100695.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P485537 | BC114974.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P485537 | BC127990.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111043 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100138 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06112745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25290649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25290607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25290658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25290655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25290650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25290660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25290623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25290617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25290647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25290653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25290629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25290611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25290621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25290628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25290612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25290631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25290614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25290625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25290640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25290627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25290611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25290623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25290631 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25290652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06591334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25290658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06472058 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25290621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25290625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25290628 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25290617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25290614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25290663 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06395184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25290627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25290647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25290662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25290612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06206237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25290645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06226937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25290640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25290644 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25290660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25290629 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06395185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25290653 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10554715 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10554717 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10554716 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10554714 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700010710.mm.1 | TC0700001214.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700010710.mm.1 | NM_146415.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010710.mm.1 | NM_146415.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010712.mm.1 | TC0700001214.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700010712.mm.1 | NM_146415.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010712.mm.1 | NM_146415.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010709.mm.1 | TC0700001214.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700010709.mm.1 | NM_146415.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010709.mm.1 | NM_146415.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 7050136 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0007050136 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2645882 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2645882 | - | - |
| mouse-6 v1.1 (Illumina) | scl0258410.1_23-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 7050136 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0258410.1_23-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7050136 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2645882 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2645882 | - | - |

Olfr291
Genomic Map