Assay Details
| Gene Symbol: Olfr742 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146430.2 | NP_666641.2 | 1 | 906 | 141 | |
| GenBank mRNA | BC132583.1 | - | 1 | 906 | 40130860 | 141 |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P116992 | NM_146430.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P116992 | BC132583.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P116992 | NM_146430.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P116992 | BC132583.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29597690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29597689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29597683 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29597705 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29597679 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11618739 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11618736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597728 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29597705 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29597716 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29597708 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11618736 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29597683 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11265902 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11618738 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29597713 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29597689 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11542132 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29597717 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11576601 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11265897 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29597690 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29597672 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11323166 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29597678 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11260115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11265900 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29597676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29597685 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29597682 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10414484 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10414483 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 901403 | - | - |
| ABI Mouse Genome Survey Microarray | 902886 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4610746 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004610746 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1249533 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1249533 | - | - |
| mouse-6 v1.1 (Illumina) | scl0258422.1_274-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4610746 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0258422.1_274-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4610746 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1249533 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1249533 | - | - |

Olfr742
Genomic Map