Assay Details
| Gene Symbol: Olfr1410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146491.1 | NP_666702.1 | 1 | 262 | 72 | |
| GenBank mRNA | BC106991.1 | - | 1 | 262 | 40044064 | 72 |
| BC106992.1 | - | 1 | 262 | 40044072 | 72 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P71624 | NM_146491.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P71624 | BC106991.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P71624 | BC106992.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P71624 | NM_146491.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P71624 | BC106991.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P71624 | BC106992.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P71624 | NM_146491.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P71624 | BC106991.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P71624 | BC106992.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P71624 | NM_146491.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P71624 | BC106991.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P71624 | BC106992.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P71624 | NM_146491.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P71624 | BC106991.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P71624 | BC106992.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P71624 | NM_146491.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P71624 | BC106991.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P71624 | BC106992.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449922 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449933 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449918 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449842 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449905 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449925 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449848 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449897 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449902 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449914 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449911 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449880 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449893 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449875 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449931 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449926 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20449916 | - | - |
| Whole Mouse Genome G4122A | A_52_P71624 | NM_146491.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P71624 | BC106991.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P71624 | BC106992.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P71624 | NM_146491.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P71624 | BC106991.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P71624 | BC106992.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120888 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00458945 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00458950 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00458948 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00458969 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00458946 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00458956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20449907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00458975 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00309932 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01146958 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01098167 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00273071 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00309935 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00458952 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01146961 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01146962 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00492555 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449912 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449916 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01146960 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449926 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449892 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01098176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449877 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449924 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449899 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449859 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449884 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01146959 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00458960 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449868 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01098175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00309933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20449853 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10348642 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10348641 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100007665.mm.1 | NM_146491.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100007665.mm.1 | NM_146491.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100007665.mm.1 | TC0100000911.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 461332 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6900017 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6900017 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006900017 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2703311 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2703311 | - | - |
| mouse-6 v1.1 (Illumina) | scl17639.1.1_77-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6900017 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl17639.1.1_77-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6900017 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2703311 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2703311 | - | - |

Olfr1410
Genomic Map