Assay Details
Gene Symbol: Olfr116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146632.1 | NP_666843.1 | 1 | 348 | 232 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2110370 | NM_146632.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147306 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2110370 | NM_146632.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169398 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169404 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169396 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169375 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169389 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169406 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169410 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169412 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169393 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169403 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169383 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169376 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169391 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169377 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169390 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169374 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169418 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169414 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169395 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169408 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169411 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169372 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169400 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31169399 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147306 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134389 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147306 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13276666 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13276664 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13192240 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13276667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169377 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169390 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169372 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169398 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169387 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169375 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169414 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169403 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31169395 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169390 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169408 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169393 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169377 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169404 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169391 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169372 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169399 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169403 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169412 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169418 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169395 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169411 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169396 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13276676 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169387 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13506685 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169414 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169400 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169374 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169375 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13192232 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169376 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169406 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169368 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169398 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31169369 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10450901 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10450900 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 401795 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 380193 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 380193 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000380193 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2682722 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2682722 | - | - |
mouse-6 v1.1 (Illumina) | scl0258625.1_9-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 380193 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0258625.1_9-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 380193 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2682722 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2682722 | - | - |
