Assay Details
| Gene Symbol: Olfr237-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146654.1 | NP_666865.1 | 1 | 344 | 159 | |
| GenBank mRNA | BC120515.1 | - | 1 | 389 | 40129438 | 159 | 
| BC120519.1 | - | 1 | 389 | 40129442 | 159 | |
 Back To Top
 Back To TopMore Information
Additional Information:
				    	 
					    	
					        	
					            	Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
					        	
					    	            
				    	
			    	
			    
			| Set Membership: |  Ambion siRNA available  Amplicon length greater than or equal to 101  G-protein coupled receptor  3' Most  Mammalian Gene Collection  Detect Genomic DNA  Recommended Assay  Mus musculus  1700  Inventoried  Both primers and probe map within a single exon | 
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
					
						
							Agilent
					
						
							Affymetrix
					
						
							ABI
					
						
							Illumina
					
					
					
						
							
								
						
					
						
							
								
						
					
						
							
								
						
					
						
							
								
						
					
				
			| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P533030 | NM_146654.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P533030 | BC120515.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P533030 | BC120519.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P533030 | NM_146654.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P533030 | BC120515.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P533030 | BC120519.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P533030 | NM_146654.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P533030 | BC120515.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P533030 | BC120519.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P533030 | NM_146654.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P533030 | BC120515.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P533030 | BC120519.1 | mouse_transcriptome | 
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P533030 | NM_146654.1 | mouse_transcriptome | 
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P533030 | BC120515.1 | mouse_transcriptome | 
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P533030 | BC120519.1 | mouse_transcriptome | 
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P533030 | NM_146654.1 | mouse_transcriptome | 
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P533030 | BC120515.1 | mouse_transcriptome | 
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P533030 | BC120519.1 | mouse_transcriptome | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24401769 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24401752 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24401771 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24401731 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24401744 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24401721 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24401740 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24401770 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24401726 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24401758 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24401734 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24401757 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24401727 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24401741 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24401754 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24401767 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24401735 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24401742 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24401738 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24401750 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24401739 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24401745 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24401755 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24401736 | - | - | 
| Whole Mouse Genome G4122A | A_52_P533030 | NM_146654.1 | mouse_transcriptome | 
| Whole Mouse Genome G4122A | A_52_P533030 | BC120515.1 | mouse_transcriptome | 
| Whole Mouse Genome G4122A | A_52_P533030 | BC120519.1 | mouse_transcriptome | 
| Whole Mouse Genome G4122A | A_52_P533030 | NM_146654.1 | mouse_transcriptome | 
| Whole Mouse Genome G4122A | A_52_P533030 | BC120515.1 | mouse_transcriptome | 
| Whole Mouse Genome G4122A | A_52_P533030 | BC120519.1 | mouse_transcriptome | 
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05669546 | - | - | 
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05668970 | - | - | 
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05668958 | - | - | 
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05669553 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401752 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401807 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401777 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401819 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401805 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401694 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401731 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401822 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401821 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401735 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401769 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401720 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401750 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401814 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401745 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401773 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401738 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401742 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401754 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401716 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401824 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401747 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401766 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401721 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401701 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401801 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401740 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401770 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401726 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401756 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401825 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401768 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401804 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401771 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401755 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401757 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24401811 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401741 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401721 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401736 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401730 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401768 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05501709 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401747 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401712 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401766 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401774 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401738 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401770 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401720 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401740 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401756 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401767 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05133795 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401726 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05596523 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05669546 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05135381 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401773 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401754 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401716 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401757 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401734 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401769 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401750 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401739 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05400668 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401735 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401727 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401755 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401758 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401777 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401752 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401745 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24401742 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05382267 | - | - | 
| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10537820 | - | - | 
| Affymetrix Mouse Gene 1.0 ST Array | 10537818 | - | - | 
| Affymetrix Mouse Gene 1.1 ST Array | 10537819 | - | - | 
| Affymetrix Mouse Gene 1.1 ST Array | 10537817 | - | - | 
| Affymetrix Mouse Clariom S Array | PSR0600003765.mm.1 | TC0600000480.mm.1 | MTA TC | 
| Affymetrix Mouse Clariom S Array | PSR0600003765.mm.1 | NM_146654.1.1 | Refseq | 
| Affymetrix Mouse Clariom S Array | PSR0600003765.mm.1 | NM_146654.1 | Refseq | 
| Affymetrix Mouse Clariom S Array | PSR0600003764.mm.1 | TC0600000480.mm.1 | MTA TC | 
| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 447482 | - | - | 
| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1170053 | - | - | 
| Illumina mouse-6 v1.0 expression beadchip | 1170053 | - | - | 
| Illumina mouseRef-8 v1.0 expression beadchip | 0001170053 | - | - | 
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1236006 | - | - | 
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1236006 | - | - | 
| mouse-6 v1.1 (Illumina) | scl0258648.1_101-S | - | - | 
| Illumina Sentrix mouse-6.v1 bead array | 1170053 | - | - | 
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0258648.1_101-S | - | - | 
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1170053 | - | - | 
| MouseRef-8 v2 Expression BeadChip | ILMN_1236006 | - | - | 
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1236006 | - | - | 
 Back To Top
 Back To Top
 Olfr237-ps1
Olfr237-ps1 Genomic Map
Genomic Map Back To Top
 Back To Top