Assay Details
| Gene Symbol: Olfr60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146955.1 | NP_667166.1 | 1 | 447 | 227 | |
| GenBank mRNA | AF102539.1 | - | 1 | 255 | 227 | |
| BC106784.1 | - | 1 | 447 | 40041376 | 227 | |
| BC106785.1 | - | 1 | 445 | 40041377 | 227 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1989640 | BC106785.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1989640 | AF102539.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1989640 | BC106784.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1989640 | NM_146955.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P180591 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1989640 | BC106785.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1989640 | AF102539.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1989640 | BC106784.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1989640 | NM_146955.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P212690 | BC106785.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P212690 | AF102539.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P212690 | BC106784.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P212690 | NM_146955.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583441 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583443 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583447 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25583422 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P180591 | - | - |
| Whole Mouse Genome G4122A | A_51_P212690 | BC106785.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P212690 | AF102539.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P212690 | BC106784.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P212690 | NM_146955.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134653 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P180591 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06424570 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06424561 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06424540 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06424554 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06424547 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06424566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25583419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25583423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06328412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25583447 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25583441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25583454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25583449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25583445 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06424554 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25583438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25583422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25583417 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25583451 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25583432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25583448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25583426 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25583430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25583452 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25583419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25583443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06424534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25583456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25583446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25583457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25583418 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25583435 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10568957 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10568956 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700037806.mm.1 | NM_146955.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700037806.mm.1 | NM_146955.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700037806.mm.1 | TC0700004498.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 812954 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1190528 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001190528 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2737550 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2737550 | - | - |
| mouse-6 v1.1 (Illumina) | scl30516.1.1_35-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1190528 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl30516.1.1_35-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1190528 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2737550 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2737550 | - | - |

Olfr60
Genomic Map