Assay Details
Gene Symbol: Olfr1243 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146969.1 | NP_667180.1 | 1 | 561 | 168 | |
GenBank mRNA | BC111812.1 | - | 1 | 556 | 168 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2117187 | NM_146969.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2117187 | BC111812.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2117187 | NM_146969.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2117187 | BC111812.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P129848 | NM_146969.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P129848 | BC111812.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P129848 | NM_146969.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P129848 | BC111812.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414057 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414025 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414028 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414055 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414049 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414073 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414058 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414020 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414042 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414071 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414074 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414048 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414067 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414087 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414031 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414065 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414084 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414022 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414054 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414072 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414064 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414014 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414059 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21414043 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414071 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414019 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414074 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414049 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414050 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414055 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414017 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414057 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414041 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414072 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414078 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414054 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414021 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414042 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414053 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414065 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414016 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414063 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414034 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414045 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414064 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414073 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414022 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414067 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414031 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01887784 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414059 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414020 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414027 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414052 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414028 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414075 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414024 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414029 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414025 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414033 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414048 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21414043 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10484829 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10484828 | - | - |
Affymetrix Mouse Clariom S Array | PSR0200034065.mm.1 | NM_146969.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200034065.mm.1 | NM_146969.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200034065.mm.1 | TC0200004062.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 898628 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 3390538 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3390538 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003390538 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2714522 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2714522 | - | - |
mouse-6 v1.1 (Illumina) | scl4275.1.1_254-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3390538 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl4275.1.1_254-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3390538 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2714522 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2714522 | - | - |
