Assay Details
Gene Symbol: Olfr668 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147059.1 | NP_667270.1 | 1 | 242 | 133 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1974932 | NM_147059.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1974932 | NM_147059.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P515949 | NM_147096.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P515949 | NM_147059.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389975 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389947 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389997 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389960 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389943 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389992 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389963 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389966 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390001 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389989 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389968 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389956 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389970 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389946 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389953 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389974 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389991 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390003 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389985 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389957 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389987 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389988 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389998 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389958 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389990 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389962 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389971 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389973 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389995 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389969 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389950 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389952 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389967 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389976 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389977 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389999 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389955 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389964 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25389951 | - | - |
Whole Mouse Genome G4122A | A_51_P515949 | NM_147096.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P515949 | NM_147059.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130323 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119308 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120300 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06144366 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06144342 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06144344 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06144369 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06144370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389975 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389970 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389887 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389895 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389980 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389966 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389884 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389992 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389969 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389974 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389962 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389905 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389947 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389989 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389977 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389882 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389953 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389889 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389909 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389960 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389990 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389967 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389973 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389978 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389998 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389904 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389894 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389997 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389985 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389897 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389958 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389957 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389900 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389987 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389952 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389963 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389950 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389910 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389955 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389906 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389982 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389896 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389976 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389968 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390003 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389964 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389979 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389991 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389999 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389980 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06305089 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389950 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389976 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389982 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389958 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389979 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389971 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389963 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389955 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389970 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389956 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389975 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05933454 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389947 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389943 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389964 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389957 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389946 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389968 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06305085 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389962 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389953 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389960 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389967 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389974 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389952 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389966 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389977 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25389978 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10566404 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10566403 | - | - |
Affymetrix Mouse Clariom S Array | PSR0700033039.mm.1 | NM_147059.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700033039.mm.1 | NM_147059.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700033039.mm.1 | TC0700003959.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 3450441 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2835637 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2835637 | - | - |
