Assay Details
| Gene Symbol: 2610044O15Rik8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153780.3 | NP_722475.1 | 2 - 3 | 264 | 80 | |
| GenBank mRNA | AK029464.1 | - | 2 - 3 | 227 | 80 | |
| AK138138.1 | - | 2 - 3 | 265 | 80 | ||
| BC023890.1 | - | 2 - 3 | 223 | 5252244 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P134785 | AK138138.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P134785 | NM_153780.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P134785 | AK011776.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P673307 | AK029464.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P673307 | AK138138.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P673307 | BC023890.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P673307 | NM_153780.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P673307 | AK029464.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P673307 | AK138138.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P673307 | BC023890.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P673307 | NM_153780.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 159603 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 240112 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 424342 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 44010 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P673307 | AK029464.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P673307 | AK138138.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P673307 | BC023890.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P673307 | NM_153780.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P673307 | AK029464.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P673307 | AK138138.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P673307 | BC023890.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P673307 | NM_153780.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P134785 | AK138138.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P134785 | NM_153780.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P134785 | AK011776.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26272455 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26272460 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26272469 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26272453 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06686239 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06686249 | - | - |
| Whole Mouse Genome G4122A | A_51_P290521 | AK138138.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P290521 | BC023890.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P290521 | NM_153780.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P290521 | AK011776.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P673307 | AK029464.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P673307 | AK138138.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P673307 | BC023890.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P673307 | NM_153780.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P673307 | AK029464.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P673307 | AK138138.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P673307 | BC023890.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P673307 | NM_153780.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119173 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134785 | AK138138.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134785 | NM_153780.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134785 | AK011776.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06686239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31456090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31456103 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13558107 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31456090 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31456064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31456092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31456099 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13393821 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31456066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31456077 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31456186 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13558108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31456103 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31456106 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31456087 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10358433 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10608651 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10453557 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452876_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428693_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10358432 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10608651 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10453555 | - | - |
| Mouse Genome 430 2.0 Array | 1452876_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1452876_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1428693_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428693_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800024749.mm.1 | NM_153780.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800024749.mm.1 | TC0800003217.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800024754.mm.1 | NM_153780.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800024754.mm.1 | TC0800003217.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800024755.mm.1 | NM_153780.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800024755.mm.1 | TC0800003217.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800024750.mm.1 | NM_153780.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800024750.mm.1 | TC0800003217.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800024751.mm.1 | NM_153780.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800024751.mm.1 | TC0800003217.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800024752.mm.1 | NM_153780.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800024752.mm.1 | TC0800003217.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800024756.mm.1 | NM_153780.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800024756.mm.1 | TC0800003217.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800024758.mm.1 | NM_153780.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800024758.mm.1 | TC0800003217.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800024753.mm.1 | NM_153780.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800024753.mm.1 | TC0800003217.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1455715_PM_at | - | - |
| Mouse Expression Set 430 | 1455715_at | - | - |
| Mouse Expression Set 430 | 1452876_PM_x_at | - | - |
| Mouse Expression Set 430 | 1428693_PM_at | - | - |
| Mouse Expression Set 430 | 1428693_at | - | - |
| Mouse Expression Set 430 | 1452876_x_at | - | - |
| Murine Genome U74v2 Set | 166856_PM_f_at | - | - |
| Murine Genome U74v2 Set | 166856_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455715_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455715_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428693_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452876_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428693_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452876_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 821062 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1940301 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1940301 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001940301 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1260441 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1260441 | - | - |
| mouse-6 v1.1 (Illumina) | scl072139.1_117-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1940301 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl072139.1_117-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1940301 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1260441 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1260441 | - | - |

2610044O15Rik8
Genomic Map