Assay Details
| Gene Symbol: Sbno2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183426.1 | NP_906271.1 | 14 - 15 | 1707 | 71 | |
| GenBank mRNA | AJ512611.1 | - | 15 - 16 | 1863 | 71 | |
| AK089496.1 | - | 10 - 11 | 1626 | 71 | ||
| AK149806.1 | - | 14 - 15 | 1731 | 71 | ||
| AK155462.1 | - | 14 - 15 | 1705 | 71 | ||
| AK155674.1 | - | 14 - 15 | 1704 | 71 | ||
| BC056369.1 | - | 14 - 15 | 1707 | 6831600 | 71 | |
| BC064113.1 | - | 11 - 12 | 1464 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sbno2
Genomic Map