Assay Details
| Gene Symbol: Foxa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291065.1 | NP_001277994.1 | 1 - 2 | 124 | 64 | |
| NM_010446.3 | NP_034576.2 | 2 - 3 | 182 | 64 | ||
| GenBank mRNA | AB231453.1 | - | 1 - 2 | 76 | 64 | |
| CF948812.1 | - | 1 - 2 | 57 | 64 | ||
| L10409.1 | - | 1 - 2 | 83 | 64 | ||
| X74937.1 | - | 2 - 3 | 182 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Citations (2)
Foxa2 programs Th2 cell-mediated innate immunity in the developing lung.
- Authors
- Chen G, Wan H, Luo F, Zhang L, Xu Y, Lewkowich I, Wills-Karp M, Whitsett JA
- Journal
- J Immunol - Page No 6133 (Tue Jun 01 00:00:00 EDT 2010)
- Volume (Year)
- 184 (2010)
- Product Usage
- -
Fibroblast growth factor 21 corrects obesity in mice.
- Authors
- Coskun T, Bina HA, Schneider MA, Dunbar JD, Hu CC, Chen Y, Moller DE, Kharitonenkov A
- Journal
- Endocrinology - Page No 6018 (Mon Dec 01 00:00:00 EST 2008)
- Volume (Year)
- 149 (2008)
- Product Usage
- -

Foxa2
Genomic Map