Assay Details
Gene Symbol: Tcp10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011553.4 | NP_035683.2 | 3 - 4 | 368 | 60 | |
NR_046291.1 | 3 - 4 | 520 | 60 | |||
GenBank mRNA | AK161455.1 | - | 3 - 4 | 339 | 60 | |
BC050771.1 | - | 3 - 4 | 334 | 6773681 | 60 | |
BC052353.1 | - | 3 - 4 | 483 | 6773668 | 60 | |
M22600.1 | - | 2 - 3 | 216 | 60 | ||
X58171.1 | - | 3 - 4 | 341 | 60 |
Gene Symbol: Tcp10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009341.2 | NP_033367.2 | 3 - 4 | 374 | 60 | |
GenBank mRNA | AK077165.1 | - | 3 - 4 | 383 | 60 | |
BC141264.1 | - | 2 - 3 | 245 | 9055876 | 60 | |
M22599.1 | - | 2 - 3 | 366 | 60 |
Gene Symbol: Tcp10c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167578.1 | NP_001161050.1 | 3 - 4 | 411 | 60 | |
GenBank mRNA | AK076614.1 | - | 3 - 4 | 357 | 60 | |
AK132653.1 | - | 3 - 4 | 409 | 60 | ||
BC061173.1 | - | 3 - 4 | 382 | 6772123 | 60 | |
CJ046593.1 | - | 3 - 4 | 409 | 60 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990815 | AK132653.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990815 | NM_001167578.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990815 | AK076614.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990815 | NM_009341.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990815 | M22602.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095583 | AK132653.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095583 | BC061173.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095583 | NM_001167578.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095583 | AK076614.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095583 | X58170.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008277 | NM_001167578.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008277 | AK076614.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P14496 | AK132653.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P14496 | NM_001167578.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P14496 | AK076614.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P14496 | AK132653.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P14496 | NM_001167578.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P14496 | AK076614.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170876 | NM_009341.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2170876 | NM_009341.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990815 | AK132653.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990815 | NM_001167578.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990815 | AK076614.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990815 | NM_009341.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990815 | M22602.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008277 | NM_001167578.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008277 | AK076614.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095583 | AK132653.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095583 | BC061173.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095583 | NM_001167578.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095583 | AK076614.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095583 | X58170.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P14496 | AK132653.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P14496 | NM_001167578.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P14496 | AK076614.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P14496 | AK132653.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P14496 | NM_001167578.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P14496 | AK076614.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P508602 | M22601.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P508602 | X58170.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03274489 | - | - |
Whole Mouse Genome G4122A | A_52_P14496 | AK132653.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P14496 | NM_001167578.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P14496 | AK076614.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P14496 | AK132653.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P14496 | NM_001167578.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P14496 | AK076614.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P508602 | M22601.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P508602 | X58170.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03274489 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13179192 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13359410 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10441836 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441590 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441844 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441595 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441841 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441829 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441591 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441831 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441593 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441597 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441593 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441599 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441854 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441833 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441592 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441593 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441595 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441835 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441828 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441841 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441852 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441838 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441837 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441839 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441852 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441598 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441825 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441852 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441825 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441838 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441596 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441836 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441599 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441832 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441590 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441598 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441834 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441600 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441839 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441833 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441597 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441594 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441837 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441832 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441846 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441833 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441846 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441844 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441834 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441594 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441835 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441831 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441598 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441589 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441827 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441830 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441839 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441840 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441838 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441831 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441829 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441840 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441827 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441834 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441846 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441592 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441591 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441826 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441828 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441832 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441830 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441589 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441830 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441595 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441825 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441590 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441589 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441840 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441599 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441592 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441827 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441829 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441597 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441837 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441826 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441594 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441600 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441591 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441600 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441836 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10441596 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1427421_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10441824 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10441824 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10441588 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10441588 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10441824 | - | - |
Mouse Genome 430 2.0 Array | 1422224_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1420527_s_at | - | - |
Mouse Genome 430 2.0 Array | 1422225_s_at | - | - |
Mouse Genome 430 2.0 Array | 1420527_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1427421_at | - | - |
Mouse Genome 430 2.0 Array | 1422224_at | - | - |
Mouse Genome 430 2.0 Array | 1422225_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1427421_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100041352_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100041352_at | - | - |
Affymetrix Murine Genome 6500A Array | Msa12110_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1700001166.mm.1 | NM_009341.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700001166.mm.1 | NM_009341.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700001166.mm.1 | TC1700002766.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700001186.mm.1 | NM_001167578.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700001186.mm.1 | NM_001167578.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700001186.mm.1 | TC1700002767.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700000565.mm.1 | NM_011553.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700000565.mm.1 | NM_011553.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700000565.mm.1 | NR_046291.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700000565.mm.1 | NR_046291.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700000565.mm.1 | TC1700000073.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700000564.mm.1 | NM_011553.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700000564.mm.1 | NM_011553.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700000564.mm.1 | NR_046291.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700000564.mm.1 | NR_046291.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700000564.mm.1 | TC1700000073.mm.1 | MTA TC |
Mouse Expression Set 430 | 1422224_PM_at | - | - |
Mouse Expression Set 430 | 1422224_at | - | - |
Mouse Expression Set 430 | 1422224_at | - | - |
Mouse Expression Set 430 | 1420527_PM_s_at | - | - |
Mouse Expression Set 430 | 1422225_s_at | - | - |
Mouse Expression Set 430 | 1420527_s_at | - | - |
Mouse Expression Set 430 | 1422224_PM_at | - | - |
Mouse Expression Set 430 | 1422224_at | - | - |
Mouse Expression Set 430 | 1422225_PM_s_at | - | - |
Mouse Expression Set 430 | 1422225_s_at | - | - |
Mouse Expression Set 430 | 1422225_PM_s_at | - | - |
Mouse Expression Set 430 | 1427421_PM_at | - | - |
Mouse Expression Set 430 | 1427421_at | - | - |
Mouse Expression Set 430 | 1422225_PM_s_at | - | - |
Mouse Expression Set 430 | 1427421_PM_at | - | - |
Mouse Expression Set 430 | 1427421_at | - | - |
Mouse Expression Set 430 | 1422224_PM_at | - | - |
Mouse Expression Set 430 | 1420527_PM_s_at | - | - |
Mouse Expression Set 430 | 1422225_s_at | - | - |
Mouse Expression Set 430 | 1420527_s_at | - | - |
Murine Genome U74v2 Set | 103058_f_at | - | - |
Murine Genome U74v2 Set | 100359_at | - | - |
Murine Genome U74v2 Set | 103058_PM_f_at | - | - |
Murine Genome U74v2 Set | 103058_f_at | - | - |
Murine Genome U74v2 Set | 100358_s_at | - | - |
Murine Genome U74v2 Set | 103058_PM_f_at | - | - |
Murine Genome U74v2 Set | 103058_f_at | - | - |
Murine Genome U74v2 Set | 103058_PM_f_at | - | - |
Murine Genome U74v2 Set | 100358_PM_s_at | - | - |
Murine Genome U74v2 Set | 100359_at | - | - |
Murine Genome U74v2 Set | 100358_s_at | - | - |
Murine Genome U74v2 Set | 100358_PM_s_at | - | - |
Murine Genome U74v2 Set | 100358_PM_s_at | - | - |
Murine Genome U74v2 Set | 100358_s_at | - | - |
Murine Genome U74v2 Set | 100359_PM_at | - | - |
Murine Genome U74v2 Set | 100359_PM_at | - | - |
Murine Genome U74v2 Set | 137662_PM_at | - | - |
Murine Genome U74v2 Set | 137662_at | - | - |
Murine Genome U74v2 Set | 137662_at | - | - |
Murine Genome U74v2 Set | 137662_PM_at | - | - |
Murine Genome U74v2 Set | 137662_at | - | - |
Murine Genome U74v2 Set | 137662_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1422224_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1422225_PM_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1422224_at | - | - |
Mouse Genome 430A 2.0 Array | 1420527_PM_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1422225_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1427421_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1427421_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1420527_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1427421_at | - | - |
Mouse Genome 430A 2.0 Array | 1422225_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1422225_PM_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1422224_at | - | - |
Mouse Genome 430A 2.0 Array | 1420527_PM_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1422224_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1422225_PM_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1422225_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1422224_at | - | - |
Mouse Genome 430A 2.0 Array | 1420527_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1427421_at | - | - |
Mouse Genome 430A 2.0 Array | 1422224_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 319225 | - | - |
ABI Mouse Genome Survey Microarray | 743997 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 5290347 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005290347 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1251610 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2451803 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2620558 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2620558 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1251610 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2451803 | - | - |
mouse-6 v1.1 (Illumina) | scl0021461.1_196-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5290347 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0021461.1_196-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5290347 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1222330 | - | - |
