Assay Details
| Gene Symbol: Akap10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019921.3 | NP_064305.2 | 7 - 8 | 1364 | 86 | |
| GenBank mRNA | AF021833.1 | - | 4 - 5 | 718 | 86 | |
| AK049399.1 | - | 7 - 8 | 1333 | 86 | ||
| AK051044.1 | - | 5 - 6 | 999 | 86 | ||
| AK083255.1 | - | 7 - 8 | 1333 | 86 | ||
| BC038483.1 | - | 4 - 5 | 422 | 86 | ||
More Information
Set Membership: |
|

Akap10
Genomic Map