Assay Details
| Gene Symbol: Nenf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025424.2 | NP_079700.1 | 3 - 4 | 368 | 61 | |
| GenBank mRNA | AB126218.1 | - | 3 - 4 | 342 | 61 | |
| AK002413.1 | - | 3 - 4 | 369 | 61 | ||
| AK004329.1 | - | 3 - 4 | 357 | 61 | ||
| AK009304.1 | - | 3 - 4 | 359 | 61 | ||
| AK020403.1 | - | 3 - 4 | 350 | 61 | ||
| AK155319.1 | - | 3 - 4 | 367 | 61 | ||
| BC048464.1 | - | 3 - 4 | 356 | 6596336 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20937432 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20937459 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20937430 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20937428 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20937431 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20937429 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1989452 | AK009304.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1989452 | AK020403.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1989452 | AK155319.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1989452 | BC048464.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1989452 | NM_025424.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 12431 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 502166 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 861044 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 274144 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 73728 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 303064 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 512871 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 175899 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117131 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168803 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1989452 | AK009304.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1989452 | AK020403.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1989452 | AK155319.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1989452 | BC048464.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1989452 | NM_025424.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P126987 | AK020403.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P126987 | BC048464.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P126987 | NM_025424.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P126987 | AK002413.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P126987 | AK004329.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20937428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20937432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20937427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20937451 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20937429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20937437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20937435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20937440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20937425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20937457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20937441 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20937444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20937452 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20937431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20937447 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20937456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20937459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20937449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20937438 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117131 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168803 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00283948 | - | - |
| Whole Mouse Genome G4122A | A_51_P126987 | AK020403.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P126987 | BC048464.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P126987 | NM_025424.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P126987 | AK002413.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P126987 | AK004329.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112544 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117131 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168803 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00523054 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00326033 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00313042 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00313043 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00857748 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00857751 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20937432 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20937431 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20937428 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20937430 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20937429 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20937459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20937347 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00843635 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00475639 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20937417 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846709 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20937352 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00412986 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01016620 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20937326 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00412982 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20937427 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20937400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20937349 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01016622 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01016618 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00067713 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00478535 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00412979 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00475641 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20937437 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20937436 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00412983 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20937440 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00704952 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20937343 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00857750 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20937347 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01016617 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20937419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20937339 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20937425 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20937444 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20937338 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20937403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00568134 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20937402 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20937433 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20937432 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00067712 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00749334 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20937341 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00568136 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00067714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00568135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00412984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00326037 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20937331 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00412988 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10361100 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10361099 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10361103 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10361102 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10361101 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417879_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10361098 | - | - |
| Mouse Genome 430 2.0 Array | 1417879_at | - | - |
| Mouse Genome 430 2.0 Array | 1417879_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66208_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66208_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100031115.mm.1 | NM_025424.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100031115.mm.1 | NM_025424.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100031115.mm.1 | TC0100003805.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100031116.mm.1 | NM_025424.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100031116.mm.1 | NM_025424.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100031116.mm.1 | TC0100003805.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100031117.mm.1 | NM_025424.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100031117.mm.1 | NM_025424.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100031117.mm.1 | TC0100003805.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100031114.mm.1 | NM_025424.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100031114.mm.1 | NM_025424.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100031114.mm.1 | TC0100003805.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417879_at | - | - |
| Mouse Expression Set 430 | 1417879_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417879_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417879_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417879_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417879_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 757052 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4210731 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004210731 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2697605 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2697605 | - | - |
| mouse-6 v1.1 (Illumina) | scl15766.4.1_27-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4210731 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl15766.4.1_27-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4210731 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2941728 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2941728 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl15766.4.1_27-S | - | - |

Nenf
Genomic Map