Assay Details
| Gene Symbol: Grap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027817.3 | NP_082093.1 | 1 - 2 | 105 | 64 | |
| GenBank mRNA | AK018457.1 | - | 1 - 2 | 106 | 64 | |
| AK153888.1 | - | 1 - 2 | 162 | 64 | ||
| BC120674.1 | - | 1 - 2 | 100 | 40129597 | 64 | |
| BC120676.1 | - | 1 - 2 | 100 | 40129599 | 64 | |
More Information
Set Membership: |
|

Grap
Genomic Map