Assay Details
| Gene Symbol: Prss27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175440.4 | NP_780649.1 | 4 - 5 | 579 | 92 | |
| GenBank mRNA | AF542056.1 | - | 4 - 5 | 515 | 92 | |
| AK080281.1 | - | 4 - 5 | 580 | 92 | ||
| BC117009.1 | - | 4 - 5 | 562 | 40126328 | 92 | |
| BC117011.1 | - | 4 - 5 | 562 | 40126330 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2007485 | AK080281.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2007485 | NM_175440.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 321704 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 646218 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 93578 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 594747 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 543631 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P134205 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2007485 | AK080281.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2007485 | NM_175440.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P263568 | AF542056.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P263568 | AK080281.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P263568 | BC117009.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P263568 | BC117011.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P263568 | NM_175440.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094930 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094955 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094964 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094958 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094966 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094959 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094961 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094947 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094931 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094924 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094926 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094962 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094953 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31094948 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P134205 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08738280 | - | - |
| Whole Mouse Genome G4122A | A_51_P263568 | AF542056.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P263568 | AK080281.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P263568 | BC117009.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P263568 | BC117011.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P263568 | NM_175440.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103895 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124427 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P134205 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13345466 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13258794 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13460722 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13345467 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13117332 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13460726 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13345464 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13460724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31095000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31094962 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094969 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094985 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094976 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094992 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094910 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094975 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13258798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31095000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13156606 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13302807 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094982 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094924 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094929 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094926 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094933 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13510312 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094922 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13341119 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094996 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094913 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094987 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094965 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094931 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094994 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094986 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094916 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13355360 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094928 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094937 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13219340 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13458374 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13258786 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094980 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13258792 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094951 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094930 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094983 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094914 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094988 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13160647 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094962 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094978 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094999 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13345471 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094958 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31094939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13510311 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10442388 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442390 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442382 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442385 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442389 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442384 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442387 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442386 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442383 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10442381 | - | - |
| Mouse Genome 430 2.0 Array | 1440686_at | - | - |
| Mouse Genome 430 2.0 Array | 1440686_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 213171_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 213171_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700002440.mm.1 | NM_175440.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002440.mm.1 | NM_175440.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002440.mm.1 | TC1700000332.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700002445.mm.1 | NM_175440.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002445.mm.1 | NM_175440.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002445.mm.1 | TC1700000332.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700002444.mm.1 | NM_175440.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002444.mm.1 | NM_175440.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002444.mm.1 | TC1700000332.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700002439.mm.1 | NM_175440.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002439.mm.1 | NM_175440.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002439.mm.1 | TC1700000332.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700002442.mm.1 | NM_175440.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002442.mm.1 | NM_175440.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002442.mm.1 | TC1700000332.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700002443.mm.1 | NM_175440.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002443.mm.1 | NM_175440.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002443.mm.1 | TC1700000332.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700002441.mm.1 | NM_175440.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002441.mm.1 | NM_175440.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002441.mm.1 | TC1700000332.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1440686_at | - | - |
| Mouse Expression Set 430 | 1440686_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 631920 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5340494 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5340494 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005340494 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2600883 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2600883 | - | - |
| mouse-6 v1.1 (Illumina) | scl51021.4.1_80-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5340494 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl51021.4.1_80-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5340494 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3059649 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2600883 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3136983 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3136983 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3059649 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2600883 | - | - |

Prss27
Genomic Map