Assay Details
| Gene Symbol: Hamp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183257.3 | NP_899080.1 | 2 - 3 | 196 | 150 | |
| GenBank mRNA | AK007975.1 | - | 2 - 3 | 186 | 150 | |
| AK008993.1 | - | 2 - 3 | 197 | 150 | ||
| AY232841.1 | - | 2 - 3 | 185 | 150 | ||
| BC086895.1 | - | 2 - 3 | 193 | 6765293 | 150 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P21486 | AK008993.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P21486 | AY232841.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P21486 | BC086895.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P21486 | NM_183257.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P21486 | AK007975.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P21486 | AK008993.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P21486 | AY232841.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P21486 | BC086895.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P21486 | NM_183257.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P21486 | AK007975.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 154367 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P180561 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P21486 | AK008993.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P21486 | AY232841.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P21486 | BC086895.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P21486 | NM_183257.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P21486 | AK007975.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P21486 | AK008993.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P21486 | AY232841.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P21486 | BC086895.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P21486 | NM_183257.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P21486 | AK007975.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P21486 | AK008993.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P21486 | AY232841.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P21486 | BC086895.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P21486 | NM_183257.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P21486 | AK007975.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P21486 | AK008993.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P21486 | AY232841.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P21486 | BC086895.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P21486 | NM_183257.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P21486 | AK007975.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034079 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034103 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034091 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034102 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034107 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P180561 | - | - |
| Whole Mouse Genome G4122A | A_52_P21486 | AK008993.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P21486 | AY232841.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P21486 | BC086895.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P21486 | NM_183257.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P21486 | AK007975.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P21486 | AK008993.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P21486 | AY232841.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P21486 | BC086895.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P21486 | NM_183257.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P21486 | AK007975.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137125 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P180561 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06085877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06085877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034086 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034080 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06085861 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06085860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034118 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034079 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10562167 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562168 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436643_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456289_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10562166 | - | - |
| Mouse Genome 430 2.0 Array | 1436643_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1436643_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66438_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66438_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700024855.mm.1 | TC0700002781.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700024855.mm.1 | NM_183257.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700024856.mm.1 | TC0700002781.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700024856.mm.1 | NM_183257.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700024854.mm.1 | TC0700002781.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700024854.mm.1 | NM_183257.4 | Refseq |
| Mouse Expression Set 430 | 1456289_at | - | - |
| Mouse Expression Set 430 | 1456289_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456289_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456289_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436643_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436643_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2940050 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2940050 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002940050 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1233659 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1233659 | - | - |
| mouse-6 v1.1 (Illumina) | scl31506.4.1_20-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2940050 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31506.4.1_20-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2940050 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2852624 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2852624 | - | - |

Hamp2
Genomic Map