Assay Details
Gene Symbol: Krtap1-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027157.3 | NP_081433.2 | 1 | 971 | 106 | |
GenBank mRNA | AI592983.1 | - | 1 | 106 | ||
AK009782.1 | - | 1 | 973 | 106 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1968799 | NM_027157.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1968799 | AK009782.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154422 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1968799 | NM_027157.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1968799 | AK009782.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28063228 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28063242 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28063226 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28063247 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28063236 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28063234 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28063257 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28063253 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28063231 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28063251 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28063249 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28063239 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28063233 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28063230 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28063245 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28063246 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28063237 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28063255 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154422 | - | - |
Whole Mouse Genome G4122A | A_51_P369773 | NM_027157.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P369773 | AK009782.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113263 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154422 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09466568 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09466538 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09466542 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09466575 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09466572 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063206 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063198 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063211 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063178 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063213 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063212 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063192 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063207 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063249 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063184 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063287 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063237 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063223 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063194 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063230 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063251 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063202 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063196 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063242 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063181 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063234 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063176 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063221 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063268 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063233 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063182 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063231 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063289 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063187 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063236 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28063216 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063198 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063211 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09466573 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063231 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063202 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09583477 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063178 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063225 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063223 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063182 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063237 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063230 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063228 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09087985 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063174 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08982424 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063251 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063224 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063206 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063253 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063207 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063203 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09066465 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063220 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09466572 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063213 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063216 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09579752 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063196 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063177 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063234 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063187 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063247 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09186436 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063257 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063221 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063218 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063205 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063200 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09466554 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063226 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063255 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063249 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063184 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063233 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09583483 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063246 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063181 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28063192 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10390900 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390899 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10390898 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1449560_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10390897 | - | - |
Mouse Genome 430 2.0 Array | 1449560_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1449560_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69664_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69664_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100034280.mm.1 | NM_027157.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100034280.mm.1 | NM_027157.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100034280.mm.1 | TC1100003716.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100034278.mm.1 | NM_027157.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100034278.mm.1 | NM_027157.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100034278.mm.1 | TC1100003716.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100034281.mm.1 | NM_027157.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100034281.mm.1 | NM_027157.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100034281.mm.1 | TC1100003716.mm.1 | MTA TC |
Mouse Expression Set 430 | 1449560_PM_at | - | - |
Mouse Expression Set 430 | 1449560_at | - | - |
Murine Genome U74v2 Set | 171147_r_at | - | - |
Murine Genome U74v2 Set | 171147_PM_r_at | - | - |
Mouse Genome 430A 2.0 Array | 1449560_at | - | - |
Mouse Genome 430A 2.0 Array | 1449560_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 479727 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 1740332 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001740332 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1257063 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1257063 | - | - |
mouse-6 v1.1 (Illumina) | scl39591.1.1_1-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1740332 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl39591.1.1_1-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1740332 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1257063 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1257063 | - | - |
