Assay Details
| Gene Symbol: Olfr223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146429.1 | NP_666640.1 | 1 | 911 | 90 | |
| GenBank mRNA | BC119106.1 | - | 1 | 1001 | 8733855 | 90 |
| BC119108.1 | - | 1 | 1001 | 8733857 | 90 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2138450 | NM_146429.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2138450 | BC119106.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2138450 | BC119108.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145684 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2138450 | NM_146429.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2138450 | BC119106.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2138450 | BC119108.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P472943 | NM_146429.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P472943 | BC119106.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P472943 | BC119108.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27837756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27837759 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27837746 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27837772 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27837758 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27837778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27837749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27837776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27837762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27837754 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27837753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27837775 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27837760 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27837742 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27837741 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27837780 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27837770 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27837744 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27837767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27837757 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27837747 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27837750 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27837768 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27837765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27837774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27837764 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145684 | - | - |
| Whole Mouse Genome G4122A | A_51_P472943 | NM_146429.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P472943 | BC119106.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P472943 | BC119108.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128328 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137699 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145684 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09060641 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09060653 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09060648 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09060649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27837749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837750 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837765 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09009573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09035212 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837758 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837790 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09173312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09723536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09602030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837728 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837747 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09217345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09724211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09060652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09606071 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27837735 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10386422 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10386421 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10386420 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100025914.mm.1 | NM_146429.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100025914.mm.1 | NM_146429.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100025914.mm.1 | TC1100002857.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100025913.mm.1 | TC1100002857.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100025915.mm.1 | TC1100002857.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6350097 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006350097 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1223993 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223993 | - | - |
| mouse-6 v1.1 (Illumina) | scl40158.1.1_2-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6350097 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl40158.1.1_2-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6350097 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1223993 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1223993 | - | - |

Olfr223
Genomic Map