Assay Details
| Gene Symbol: Olfr1276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146395.1 | NP_666507.1 | 1 | 612 | 141 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2101671 | NM_146395.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2025340 | NM_146395.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177553 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2025340 | NM_146395.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2101671 | NM_146395.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P192325 | NM_146395.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524585 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524593 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524620 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524604 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524590 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524617 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524582 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524581 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524598 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524599 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524602 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524592 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524579 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524623 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524580 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524596 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524608 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524587 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524613 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524591 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524588 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524600 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524586 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21524577 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177553 | - | - |
| Whole Mouse Genome G4122A | A_52_P192325 | NM_146395.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106939 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112911 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177553 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02209426 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02209421 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01941480 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02209434 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02209429 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01941488 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02209433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21524640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21524608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21524606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21524610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21524642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21524591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21524623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21524580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21524599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21524596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21524593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21524585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21524588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21524592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21524587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21524621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21524581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21524586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21524604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21524590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21524579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21524577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21524600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21524598 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21524621 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21524590 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21524602 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21524599 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21524596 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21524626 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21524580 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21524633 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21524586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21524617 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21524579 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21524587 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01709645 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01941474 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02209418 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21524631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02209433 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21524577 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01343130 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21524582 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01180786 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21524604 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21524600 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21524574 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21524593 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21524608 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21524585 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02209421 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21524606 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21524613 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21524592 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01624700 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21524591 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21524581 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01180785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21524598 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10474480 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10474479 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200012257.mm.1 | NM_146395.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200012257.mm.1 | NM_146395.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200012257.mm.1 | TC0200001639.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 751708 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 520215 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 520215 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000520215 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1231917 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1231917 | - | - |
| mouse-6 v1.1 (Illumina) | scl20497.1.1_55-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 520215 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl20497.1.1_55-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 520215 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1231917 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1231917 | - | - |

Olfr1276
Genomic Map