Assay Details
Gene Symbol: Olfr126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146890.2 | NP_667101.1 | 1 | 259 | 130 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2150238 | NM_146890.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2150238 | NM_146890.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P149531 | NM_146890.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31170220 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31170196 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31170193 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31170200 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31170175 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31170178 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31170182 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31170218 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31170189 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31170183 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31170217 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31170174 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31170198 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31170176 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31170201 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31170180 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31170187 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31170202 | - | - |
Whole Mouse Genome G4122A | A_51_P149531 | NM_146890.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122500 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115654 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118680 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101378 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13224852 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13433993 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13224848 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170194 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170173 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170234 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170196 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170171 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170182 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170192 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170187 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170230 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170155 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170237 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170166 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170180 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170164 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170175 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170231 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170157 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170172 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170163 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170236 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170198 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170169 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31170220 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170177 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170160 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170217 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170228 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170163 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170186 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170230 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170169 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170180 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170236 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13224848 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170157 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170218 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170187 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170196 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13389698 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170231 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170175 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170158 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170198 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170155 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170162 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170200 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170168 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13433991 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13224853 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170176 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170220 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170234 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170189 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170178 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170183 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170170 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170235 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170237 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170179 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170166 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170224 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170192 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170164 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13433998 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170174 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170161 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13438334 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170182 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31170216 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10450907 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10445170 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10445166 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10445169 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10445165 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10450906 | - | - |
Affymetrix Mouse Clariom S Array | PSR1700007749.mm.1 | NM_146890.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700007749.mm.1 | NM_146890.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700007749.mm.1 | TC1700000787.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 6350035 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 840348 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000840348 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006350035 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2775789 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1238397 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1238397 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2775789 | - | - |
mouse-6 v1.1 (Illumina) | scl00258892.1_319-S | - | - |
mouse-6 v1.1 (Illumina) | scl0258892.1_320-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6350035 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 840348 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0258892.1_320-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl00258892.1_319-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6350035 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 840348 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2775788 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1238397 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1238397 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2775788 | - | - |
