Assay Details
Gene Symbol: Olfr295 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146851.2 | NP_667062.2 | 1 | 355 | 149 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2065475 | NM_146851.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2065475 | NM_146851.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P183401 | NM_146851.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25295190 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25295189 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25295173 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25295187 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25295200 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25295183 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25295174 | - | - |
Whole Mouse Genome G4122A | A_51_P183401 | NM_146851.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123627 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110417 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06047778 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06047794 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25295152 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25295221 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25295222 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25295169 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25295217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25295174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25295218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25295183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25295189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25295224 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25295215 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25295168 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25295219 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25295205 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06226398 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06060820 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25295168 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25295212 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25295172 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25295169 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25295153 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06047794 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25295156 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05950104 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25295174 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25295200 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25295189 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25295161 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25295183 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05950112 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10554730 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10554729 | - | - |
Affymetrix Mouse Clariom S Array | PSR0700010775.mm.1 | TC0700001244.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700010775.mm.1 | NM_146851.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700010775.mm.1 | NM_146851.2 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 893218 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 7040524 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 7040524 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0007040524 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1225728 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1225728 | - | - |
mouse-6 v1.1 (Illumina) | scl0258850.1_189-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 7040524 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0258850.1_189-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7040524 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1225728 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1225728 | - | - |
