Assay Details
| Gene Symbol: Wibg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170869.2 | NP_001164340.1 | 3 | 350 | 150 | |
| NM_001253704.1 | NP_001240633.1 | 4 | 405 | 150 | ||
| NM_001253705.1 | NP_001240634.1 | 3 | 509 | 150 | ||
| NM_030100.4 | NP_084376.2 | 3 | 358 | 150 | ||
| GenBank mRNA | AJ459407.1 | - | 3 | 275 | 150 | |
| AK020702.1 | - | 3 | 349 | 150 | ||
| BC049647.1 | - | 3 | 283 | 6707378 | 150 | |
| BI989958.1 | - | 3 | 350 | 150 | ||
| BQ177652.1 | - | 3 | 277 | 150 | ||
| CK332572.1 | - | 3 | 299 | 150 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27543032 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27543036 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27543035 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27543034 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27543031 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27543029 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27543030 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2100997 | AJ459407.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2100997 | AK020702.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 440858 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478342 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 300304 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 7579 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 716527 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 409837 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 678483 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 612299 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 823600 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 868961 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 675858 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 882415 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 54802 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 160022 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124533 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136049 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2100997 | AJ459407.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2100997 | AK020702.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543033 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543043 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543037 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543030 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543040 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27543047 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07234076 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P124533 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136049 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02247865 | - | - |
| Whole Mouse Genome G4122A | A_52_P298071 | CK332572.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P298071 | NR_033208.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P298071 | AJ459407.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P298071 | AK020702.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P298071 | BC049647.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140568 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136049 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P124533 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08672888 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08672907 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08672906 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27543031 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27543030 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27543034 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27543032 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27543036 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27543029 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27543035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27543106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543057 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08734129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08641197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08715443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08798363 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543033 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543038 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543119 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08436072 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543091 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08358677 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08715444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08711959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543020 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543066 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543025 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543116 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543151 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543117 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08436076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543045 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08762740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08763421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543048 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08436073 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543149 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08641191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543032 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543062 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08761521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543064 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08715330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08436726 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543023 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08641189 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543143 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543067 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543112 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543037 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543055 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543043 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543061 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543053 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543022 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27543087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08641208 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10367397 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10367399 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10367398 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10367395 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10367396 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453315_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10367393 | - | - |
| Mouse Genome 430 2.0 Array | 1453315_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453315_at | - | - |
| Mouse Expression Set 430 | 1453315_PM_at | - | - |
| Mouse Expression Set 430 | 1453315_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453315_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453315_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 670633 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 102480484 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102480484 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2230088 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002230088 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2665791 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2665791 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1252682 | - | - |
| mouse-6 v1.1 (Illumina) | scl38272.5_429-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0078428.2_263-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2230088 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102480484 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0078428.2_263-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2230088 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102480484 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1252682 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1252682 | - | - |

Wibg
Genomic Map