Assay Details
| Gene Symbol: Rps13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026533.3 | NP_080809.1 | 4 | 318 | 111 | |
| GenBank mRNA | AK012479.1 | - | 4 | 263 | 111 | |
| AK018703.1 | - | 4 | 262 | 111 | ||
| AK076060.1 | - | 4 | 262 | 111 | ||
| AK088237.1 | - | 3 | 895 | 111 | ||
| AK151043.1 | - | 4 | 263 | 111 | ||
| AK151671.1 | - | 4 | 264 | 111 | ||
| AK161798.1 | - | 3 | 861 | 111 | ||
| AK166691.1 | - | 4 | 263 | 111 | ||
| AK177445.1 | - | 261 | 111 | |||
| AV498110.1 | - | 3 | 225 | 111 | ||
| BC011192.1 | - | 4 | 261 | 4016679 | 111 | |
| BC085269.1 | - | 2 | 967 | 111 | ||
| BC090397.1 | - | 4 | 245 | 5150054 | 111 | |
| BC106137.1 | - | 4 | 246 | 4990874 | 111 | |
| BC125602.1 | - | 4 | 262 | 40130117 | 111 | |
| BC125604.1 | - | 4 | 262 | 40130119 | 111 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25447592 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25447596 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25447597 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25447591 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25447594 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25447595 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25447599 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2146749 | BC106137.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2146749 | BC125602.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2146749 | BC125604.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2146749 | AK088237.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2146749 | AK151043.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963084 | BC011192.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963084 | BC090397.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963084 | BC106137.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963084 | AV498110.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963084 | AK088237.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 742938 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 32757 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171360 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2146749 | BC106137.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2146749 | BC125602.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2146749 | BC125604.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2146749 | AK088237.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2146749 | AK151043.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963084 | BC011192.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963084 | BC090397.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963084 | BC106137.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963084 | AV498110.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963084 | AK088237.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P327469 | BC106137.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P327469 | AK088237.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P327469 | AK151043.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P327469 | AK151671.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P327469 | AK161798.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P646112 | BC011192.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447612 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447582 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447587 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447572 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447598 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447594 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447605 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447592 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447585 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447580 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447617 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447614 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447588 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447616 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447604 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447624 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447620 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447623 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447601 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447607 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447574 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447599 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447630 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447586 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447583 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447627 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447596 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447622 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447578 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447626 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447591 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447568 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447603 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25447619 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171360 | - | - |
| Whole Mouse Genome G4122A | A_51_P327469 | BC106137.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P327469 | AK088237.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P327469 | AK151043.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P327469 | AK151671.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P327469 | AK161798.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P646112 | BC011192.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139755 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171360 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05978377 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06242916 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05971357 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06242922 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06242929 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06242923 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06242917 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06242920 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06242913 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25447596 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25447599 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25447597 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25447595 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25447592 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25447591 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25447594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25447598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447601 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447586 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447568 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447554 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06555209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447596 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06295458 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447591 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447553 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06067332 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06015011 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447583 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06512258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06242918 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06067327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447574 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06298271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447576 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447585 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447588 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447563 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447587 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447549 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06500089 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447604 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447592 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447582 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25447567 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10567166 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567172 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567170 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567167 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10565436 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567169 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10565435 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573694 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567168 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567165 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573695 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567164 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10567163 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10567171 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10573693 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10565434 | - | - |
| Mouse Genome 430 2.0 Array | 1448142_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1415913_at | - | - |
| Mouse Genome 430 2.0 Array | 1438794_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1415912_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1415912_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1448142_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1415913_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438794_PM_x_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700034469.mm.1 | NM_026533.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700034469.mm.1 | NM_026533.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700034469.mm.1 | TC0700004155.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700034468.mm.1 | NM_026533.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700034468.mm.1 | NM_026533.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700034468.mm.1 | TC0700004155.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700034466.mm.1 | TC0700004155.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700034467.mm.1 | TC0700004155.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1415913_PM_at | - | - |
| Mouse Expression Set 430 | 1415912_a_at | - | - |
| Mouse Expression Set 430 | 1448142_PM_x_at | - | - |
| Mouse Expression Set 430 | 1415912_PM_a_at | - | - |
| Mouse Expression Set 430 | 1438794_PM_x_at | - | - |
| Mouse Expression Set 430 | 1415913_at | - | - |
| Mouse Expression Set 430 | 1448142_x_at | - | - |
| Mouse Expression Set 430 | 1438794_x_at | - | - |
| Murine Genome U74v2 Set | 93027_r_at | - | - |
| Murine Genome U74v2 Set | 93027_PM_r_at | - | - |
| Murine Genome U74v2 Set | 134043_at | - | - |
| Murine Genome U74v2 Set | 134043_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415913_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438794_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415912_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438794_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415912_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448142_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415913_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448142_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 605820 | - | - |
| ABI Mouse Genome Survey Microarray | 902457 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1570364 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102970711 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1570364 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001570364 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2482591 | - | - |
| mouse-6 v1.1 (Illumina) | scl068052.3_30-S | - | - |
| mouse-6 v1.1 (Illumina) | scl30771.2_110-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1570364 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102970711 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl068052.3_30-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102970711 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1570364 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2940017 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2482591 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2940017 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1222723 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl068052.3_30-S | - | - |

Rps13
Genomic Map