Assay Details
| Gene Symbol: Fam136a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025591.2 | NP_079867.1 | 3 | 1263 | 154 | |
| GenBank mRNA | AK005280.1 | - | 3 | 1223 | 154 | |
| AK141279.1 | - | 3 | 1223 | 154 | ||
| BC024049.1 | - | 3 | 1263 | 5100000 | 154 | |
| BC103775.1 | - | 3 | 1264 | 6485116 | 154 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24609732 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24609731 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24609727 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24609728 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24609733 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054927 | AK141279.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054927 | BC024049.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054927 | BC103775.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054927 | NM_025591.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054927 | AK005280.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 374841 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 152844 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054927 | AK141279.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054927 | BC024049.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054927 | BC103775.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054927 | NM_025591.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054927 | AK005280.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609700 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609719 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609713 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609740 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609710 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609724 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609711 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609723 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609735 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609716 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609721 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609742 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609708 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609730 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609718 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609699 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609734 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609728 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609707 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609744 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609722 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609704 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609738 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609739 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24609714 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01375391 | - | - |
| Whole Mouse Genome G4122A | A_52_P110002 | AK141279.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P110002 | BC024049.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P110002 | BC103775.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P110002 | NM_025591.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P110002 | AK005280.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106448 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138020 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05087474 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05087473 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05430442 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24609733 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24609727 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24609732 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24609731 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24609728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609691 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609687 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05386767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05849546 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609683 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609686 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05635513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05635514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05087473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609713 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609735 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05804231 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05323893 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05559511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609734 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05804229 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609698 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05087474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609723 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05386770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609716 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609702 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609721 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05635515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609719 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10539703 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539704 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1457611_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1441796_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417075_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10539702 | - | - |
| Mouse Genome 430 2.0 Array | 1417075_at | - | - |
| Mouse Genome 430 2.0 Array | 1441796_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417075_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1457611_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1441796_at | - | - |
| Mouse Genome 430 2.0 Array | 1457611_PM_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66488_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66488_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600007595.mm.1 | TC0600000985.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600007595.mm.1 | NM_025591.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600007595.mm.1 | NM_025591.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600007593.mm.1 | TC0600000985.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600007593.mm.1 | NM_025591.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600007593.mm.1 | NM_025591.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600007594.mm.1 | TC0600000985.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600007594.mm.1 | NM_025591.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600007594.mm.1 | NM_025591.2 | Refseq |
| Mouse Expression Set 430 | 1417075_PM_at | - | - |
| Mouse Expression Set 430 | 1417075_at | - | - |
| Mouse Expression Set 430 | 1457611_x_at | - | - |
| Mouse Expression Set 430 | 1441796_PM_at | - | - |
| Mouse Expression Set 430 | 1441796_at | - | - |
| Mouse Expression Set 430 | 1457611_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417075_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417075_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417075_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417075_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457611_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441796_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457611_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441796_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 898112 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 110050 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000110050 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2642188 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2642188 | - | - |
| mouse-6 v1.1 (Illumina) | scl0066488.2_231-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 110050 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0066488.2_231-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 110050 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2642188 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2642188 | - | - |

Fam136a
Genomic Map