Assay Details
| Gene Symbol: Tma7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183250.2 | NP_899073.1 | 3 | 182 | 119 | |
| GenBank mRNA | AK003762.1 | - | 3 | 183 | 119 | |
| AK160705.1 | - | 3 | 180 | 119 | ||
| AK169860.1 | - | 3 | 176 | 119 | ||
| BC034602.1 | - | 3 | 164 | 119 | ||
| BC054363.1 | - | 3 | 155 | 119 | ||
| BC090400.1 | - | 3 | 155 | 119 | ||
| BC106104.1 | - | 3 | 154 | 5029265 | 119 | |
| BC116987.1 | - | 3 | 178 | 40126306 | 119 | |
| BC116989.1 | - | 3 | 178 | 40126308 | 119 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26818178 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26818180 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26818181 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26818182 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2100864 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999548 | BC054363.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999548 | NM_183250.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999548 | AK003762.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 148269 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 143068 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 114535 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 105138 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178300 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2100864 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999548 | BC054363.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999548 | NM_183250.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999548 | AK003762.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818178 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818213 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818193 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818187 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818195 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818201 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818208 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818207 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818171 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818186 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818184 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818197 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818206 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818192 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26818167 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178300 | - | - |
| Whole Mouse Genome G4122A | A_52_P303041 | BC054363.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P303041 | BC090400.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P303041 | BC106104.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P303041 | BC034602.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P303041 | NM_183250.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P431199 | BC054363.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P431199 | NM_183250.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P431199 | AK003762.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P431199 | AK169860.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139277 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178300 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07498687 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07878106 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07498688 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07878101 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26818181 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26818178 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26818180 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26818182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26818223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26818193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08030451 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26818175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26818173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26818177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26818190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26818187 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26818171 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26818164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26818188 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26818167 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08079167 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26818184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26818182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08079173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26818189 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08097063 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08030453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26818186 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10351666 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10596985 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10596987 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10596984 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10596986 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10596983 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454813_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10596982 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10351665 | - | - |
| Mouse Genome 430 2.0 Array | 1454813_at | - | - |
| Mouse Genome 430 2.0 Array | 1454813_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900025915.mm.1 | NM_183250.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900025915.mm.1 | NM_183250.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900025915.mm.1 | TC0900003074.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900025917.mm.1 | NM_183250.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900025917.mm.1 | NM_183250.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900025917.mm.1 | TC0900003074.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900025919.mm.1 | NM_183250.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900025919.mm.1 | NM_183250.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900025919.mm.1 | TC0900003074.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900025920.mm.1 | NM_183250.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900025920.mm.1 | NM_183250.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900025920.mm.1 | TC0900003074.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900025916.mm.1 | NM_183250.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900025916.mm.1 | NM_183250.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900025916.mm.1 | TC0900003074.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900025918.mm.1 | NM_183250.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900025918.mm.1 | NM_183250.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900025918.mm.1 | TC0900003074.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1454813_at | - | - |
| Mouse Expression Set 430 | 1454813_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1454813_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1454813_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1454813_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1454813_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 895948 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2360673 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2360673 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002360673 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2669699 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2669699 | - | - |
| mouse-6 v1.1 (Illumina) | scl066167.1_147-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2360673 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl066167.1_147-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2360673 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2669699 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2669699 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl066167.1_147-S | - | - |

Tma7
Genomic Map