Assay Details
| Gene Symbol: Hdac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008228.2 | NP_032254.1 | 8 - 9 | 859 | 67 | |
| GenBank mRNA | BC092070.1 | - | 8 - 9 | 854 | 4217199 | 67 |
| BC092285.1 | - | 8 - 9 | 859 | 67 | ||
| BC108371.1 | - | 8 - 9 | 860 | 4976514 | 67 | |
| U80780.1 | - | 8 - 9 | 856 | 67 | ||
| X98207.1 | - | 8 - 9 | 859 | 67 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hdac1
Genomic Map