Assay Details
| Gene Symbol: Ensa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026212.1 | NP_001021383.1 | 2 | 294 | 144 | |
| NM_019561.2 | NP_062507.1 | 2 | 294 | 144 | ||
| GenBank mRNA | AJ005985.1 | - | 2 | 154 | 144 | |
| AK006149.1 | - | 2 | 264 | 144 | ||
| AK149726.1 | - | 2 | 262 | 144 | ||
| AK161290.1 | - | 2 | 260 | 144 | ||
| BC058802.1 | - | 2 | 243 | 6475839 | 144 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957419 | NM_019561.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957419 | AK006149.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P100620 | BC020533.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P100620 | BC058802.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P100620 | AK161290.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P100620 | NM_001026212.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 768961 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 505041 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 663588 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 458975 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 174447 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957419 | NM_019561.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957419 | AK006149.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P100620 | BC020533.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P100620 | BC058802.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P100620 | AK161290.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P100620 | NM_001026212.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P513600 | BC020533.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P513600 | BC058802.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P513600 | AK149726.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P513600 | AK161290.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P513600 | NM_001026212.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P222021 | AJ005985.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P145076 | NM_019561.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P145076 | AI451143.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P145076 | AK006149.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22344770 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22344756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22344758 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22344774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22344788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22344753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22344779 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22344773 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22344785 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22344750 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22344778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22344789 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22344780 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22344782 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22344777 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22344768 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22344762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22344752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22344760 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22344755 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04995558 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00702868 | - | - |
| Whole Mouse Genome G4122A | A_52_P222021 | AJ005985.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P513600 | BC020533.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P513600 | BC058802.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P513600 | AK149726.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P513600 | AK161290.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P513600 | NM_001026212.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P145076 | NM_019561.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P145076 | AI451143.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P145076 | AK006149.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110725 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100620 | BC020533.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100620 | BC058802.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100620 | AK161290.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100620 | NM_001026212.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00702868 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02911127 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02911129 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02911132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22344821 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344770 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344758 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344788 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344823 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02911132 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344782 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344813 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02848360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344822 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344806 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344819 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344798 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344827 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344762 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344752 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344821 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344826 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344790 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344792 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344771 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02506660 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344761 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344768 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344796 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03191235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344760 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02951942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02731961 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344756 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344825 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344753 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344777 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344808 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344830 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344750 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03189399 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344778 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344769 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22344780 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02553655 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10494303 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494305 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494304 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494302 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494300 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494301 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423330_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428620_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10494299 | - | - |
| Mouse Genome 430 2.0 Array | 1428620_at | - | - |
| Mouse Genome 430 2.0 Array | 1428620_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423330_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423330_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56205_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56205_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300007237.mm.1 | NM_001026212.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300007237.mm.1 | NM_001026212.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300007237.mm.1 | TC0300000908.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300007236.mm.1 | NM_001026212.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300007236.mm.1 | NM_001026212.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300007236.mm.1 | NM_019561.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300007236.mm.1 | NM_019561.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300007236.mm.1 | TC0300000908.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428620_PM_at | - | - |
| Mouse Expression Set 430 | 1428620_at | - | - |
| Mouse Expression Set 430 | 1423330_PM_at | - | - |
| Mouse Expression Set 430 | 1423330_at | - | - |
| Murine Genome U74v2 Set | 93722_PM_at | - | - |
| Murine Genome U74v2 Set | 93722_at | - | - |
| Murine Genome U74v2 Set | 167271_i_at | - | - |
| Murine Genome U74v2 Set | 167271_PM_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423330_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428620_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423330_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428620_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423330_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423330_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428620_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428620_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 540821 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 105290465 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6620546 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006620546 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2763294 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2763294 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2440407 | - | - |
| mouse-6 v1.1 (Illumina) | scl22877.5_401-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0056205.2_264-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6620546 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105290465 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0056205.2_264-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105290465 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6620546 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3112185 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3032778 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3032778 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3112185 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2763294 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2440407 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0056205.2_264-S | - | - |

Ensa
Genomic Map