Assay Details
| Gene Symbol: Mtf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253877.1 | NP_001240806.1 | 10 - 11 | 1573 | 161 | |
| NM_001253878.1 | NP_001240807.1 | 10 - 11 | 1292 | 161 | ||
| NM_001253879.1 | NP_001240808.1 | 9 - 10 | 1215 | 161 | ||
| NM_001253880.1 | NP_001240809.1 | 7 - 8 | 930 | 161 | ||
| NM_013827.3 | NP_038855.2 | 9 - 10 | 1211 | 161 | ||
| NR_045608.1 | 11 - 12 | 1279 | 161 | |||
| GenBank mRNA | AB221622.1 | - | 6 - 7 | 618 | 161 | |
| AK030752.1 | - | 9 - 10 | 1161 | 161 | ||
| AK031409.1 | - | 11 - 12 | 1280 | 161 | ||
| AK032798.1 | - | 10 - 11 | 1534 | 161 | ||
| AK032819.1 | - | 10 - 11 | 1535 | 161 | ||
| AK137539.1 | - | 11 - 12 | 1520 | 161 | ||
| AK139685.1 | - | 10 - 11 | 1293 | 161 | ||
| BC050826.1 | - | 9 - 10 | 1165 | 161 | ||
| BC057163.1 | - | 9 - 10 | 1008 | 161 | ||
| BC071223.1 | - | 7 - 8 | 683 | 161 | ||
| BC092237.1 | - | 9 - 10 | 1143 | 30637248 | 161 | |
| BC100340.1 | - | 7 - 8 | 861 | 30715034 | 161 | |
| S78454.1 | - | 6 - 7 | 615 | 158 | ||
More Information
Set Membership: |
|

Mtf2
Genomic Map