Assay Details
| Gene Symbol: Akt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011785.3 | NP_035915.3 | 1 - 2 | 86 | 81 | |
| GenBank mRNA | AF124142.1 | - | 1 - 2 | 76 | 81 | |
| BC066861.1 | - | 1 - 2 | 86 | 30089997 | 81 | |
| CK390299.1 | - | 2 - 3 | 255 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Akt3
Genomic Map