Assay Details
| Gene Symbol: Adnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009628.2 | NP_033758.2 | 2 - 3 | 590 | 144 | |
| GenBank mRNA | AK129214.1 | - | 2 - 3 | 641 | 145 | |
| AK134786.1 | - | 1 - 2 | 112 | 144 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adnp
Genomic Map