Assay Details
Gene Symbol: Smo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176996.4 | NP_795970.3 | 7 - 8 | 1883 | 64 | |
GenBank mRNA | AK081050.1 | - | 1 - 2 | 71 | 64 | |
AK193396.1 | - | 3 - 4 | 270 | 64 | ||
BC096028.1 | - | 7 - 8 | 1402 | 6850065 | 64 | |
BQ947565.1 | - | 2 - 3 | 145 | 64 | ||
CA749812.1 | - | 4 - 5 | 483 | 64 | ||
FJ883552.1 | - | 7 - 8 | 1392 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
