Assay Details
Gene Symbol: Bcl11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021399.2 | NP_067374.2 | 2 - 3 | 697 | 66 | |
GenBank mRNA | AB043553.1 | - | 2 - 3 | 697 | 66 | |
BC019503.1 | - | 2 - 3 | 497 | 4457123 | 66 | |
BQ745012.1 | - | 1 - 2 | 312 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
