Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900020278.mm.1 |
NM_019927.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020278.mm.1 |
NM_019927.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020278.mm.1 |
TC0900002406.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900020275.mm.1 |
NM_019927.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020275.mm.1 |
NM_019927.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020275.mm.1 |
TC0900002406.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900020277.mm.1 |
NM_019927.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020277.mm.1 |
NM_019927.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020277.mm.1 |
TC0900002406.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900020280.mm.1 |
NM_019927.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020280.mm.1 |
NM_019927.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020280.mm.1 |
TC0900002406.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900020271.mm.1 |
NM_019927.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020271.mm.1 |
NM_019927.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020271.mm.1 |
TC0900002406.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900020274.mm.1 |
NM_019927.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020274.mm.1 |
NM_019927.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020274.mm.1 |
TC0900002406.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900020281.mm.1 |
TC0900002406.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900020276.mm.1 |
NM_019927.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020276.mm.1 |
NM_019927.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020276.mm.1 |
TC0900002406.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900020279.mm.1 |
NM_019927.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020279.mm.1 |
NM_019927.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020279.mm.1 |
TC0900002406.mm.1 |
MTA TC |