Assay Details
Gene Symbol: Capza2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007604.2 | NP_031630.1 | 6 - 7 | 528 | 90 | |
GenBank mRNA | AK007550.1 | - | 6 - 7 | 527 | 90 | |
AK008204.1 | - | 6 - 7 | 514 | 90 | ||
AK150587.1 | - | 6 - 7 | 530 | 90 | ||
AK150751.1 | - | 6 - 7 | 595 | 90 | ||
AK152674.1 | - | 3 - 4 | 322 | 90 | ||
AY544984.1 | - | 6 - 7 | 524 | 90 | ||
BC082589.1 | - | 6 - 7 | 528 | 30626416 | 90 | |
U16741.1 | - | 6 - 7 | 609 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
