Assay Details
| Gene Symbol: St7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083315.2 | NP_001076784.1 | 9 - 10 | 1297 | 80 | |
| NM_001289624.1 | NP_001276553.1 | 10 - 11 | 1290 | 80 | ||
| NM_001289625.1 | NP_001276554.1 | 10 - 11 | 1290 | 80 | ||
| NM_001289626.1 | NP_001276555.1 | 9 - 10 | 1221 | 80 | ||
| NM_001289627.1 | NP_001276556.1 | 10 - 11 | 1024 | 80 | ||
| NM_001289629.1 | NP_001276558.1 | 9 - 10 | 955 | 80 | ||
| NM_022332.3 | NP_071727.2 | 10 - 11 | 1366 | 80 | ||
| GenBank mRNA | AK079107.1 | - | 10 - 11 | 1367 | 80 | |
| AK132103.1 | - | 9 - 10 | 1251 | 80 | ||
| AK151550.1 | - | 9 - 10 | 1252 | 80 | ||
| AY007792.1 | - | 10 - 11 | 1252 | 80 | ||
| AY007793.1 | - | 9 - 10 | 1183 | 80 | ||
| AY007794.1 | - | 11 - 12 | 1281 | 80 | ||
| AY007795.1 | - | 10 - 11 | 1212 | 80 | ||
| AY007796.1 | - | 11 - 12 | 1091 | 80 | ||
| AY007797.1 | - | 10 - 11 | 1022 | 80 | ||
| AY007798.1 | - | 10 - 11 | 1024 | 80 | ||
| AY007799.1 | - | 9 - 10 | 955 | 80 | ||
| AY551821.1 | - | 10 - 11 | 1017 | 80 | ||
| AY551822.1 | - | 9 - 10 | 948 | 80 | ||
| BC012719.1 | - | 4 - 5 | 437 | 80 | ||
| BC024652.1 | - | 10 - 11 | 1290 | 3660309 | 80 | |
| BC051429.1 | - | 9 - 10 | 2976 | 80 | ||
| BC060630.1 | - | 9 - 10 | 3039 | 80 | ||
| BC108418.1 | - | 10 - 11 | 1310 | 30603484 | 80 | |
| CT010245.1 | - | 9 - 10 | 874 | 80 | ||
More Information
Set Membership: |
|

St7
Genomic Map