Assay Details
| Gene Symbol: Calcr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042725.1 | NP_001036190.1 | 11 - 12 | 1220 | 68 | |
| NM_007588.2 | NP_031614.2 | 12 - 13 | 1580 | 68 | ||
| GenBank mRNA | AF056329.1 | - | 3 - 4 | 260 | 68 | |
| AK039161.1 | - | 12 - 13 | 1525 | 68 | ||
| AK133983.1 | - | 11 - 12 | 1221 | 68 | ||
| AK138275.1 | - | 11 - 12 | 1471 | 68 | ||
| AK162391.1 | - | 12 - 13 | 1335 | 68 | ||
| BC119232.1 | - | 10 - 11 | 1087 | 8733981 | 68 | |
| BC119272.1 | - | 10 - 11 | 1087 | 8734021 | 68 | |
| U18542.1 | - | 12 - 13 | 1482 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Calcr
Genomic Map