Assay Details
| Gene Symbol: Calcr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042725.1 | NP_001036190.1 | 13 - 14 | 1508 | 103 | |
| NM_007588.2 | NP_031614.2 | 14 - 15 | 1868 | 103 | ||
| GenBank mRNA | AK039161.1 | - | 14 - 15 | 1813 | 103 | |
| AK133983.1 | - | 13 - 14 | 1509 | 103 | ||
| AK138275.1 | - | 13 - 14 | 1759 | 103 | ||
| AK162391.1 | - | 14 - 15 | 1623 | 103 | ||
| BC119232.1 | - | 12 - 13 | 1375 | 8733981 | 103 | |
| BC119272.1 | - | 12 - 13 | 1375 | 8734021 | 103 | |
| U18542.1 | - | 14 - 15 | 1770 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Calcr
Genomic Map