Assay Details
Gene Symbol: Calcr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042725.1 | NP_001036190.1 | 2 - 3 | 224 | 67 | |
NM_007588.2 | NP_031614.2 | 2 - 3 | 473 | 67 | ||
GenBank mRNA | AK039161.1 | - | 2 - 3 | 418 | 67 | |
AK133983.1 | - | 2 - 3 | 225 | 67 | ||
AK138275.1 | - | 2 - 3 | 475 | 67 | ||
AK162391.1 | - | 2 - 3 | 228 | 67 | ||
CJ057218.1 | - | 2 - 3 | 474 | 67 | ||
U18542.1 | - | 2 - 3 | 375 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK039161.1, AK133983.1, AK138275.1, AK162391.1, NM_001042725.1, NM_007588.2, U18542.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
