Assay Details
| Gene Symbol: Mboat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153546.4 | NP_705774.1 | 2 - 3 | 421 | 76 | |
| GenBank mRNA | AB297382.1 | - | 2 - 3 | 244 | 76 | |
| AB305045.1 | - | 2 - 3 | 244 | 76 | ||
| AK033388.1 | - | 2 - 3 | 363 | 76 | ||
| AK137755.1 | - | 2 - 3 | 353 | 76 | ||
| AK146984.1 | - | 2 - 3 | 328 | 76 | ||
| BC023845.1 | - | 2 - 3 | 425 | 5346000 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mboat1
Genomic Map