Assay Details
| Gene Symbol: Mboat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153546.4 | NP_705774.1 | 3 - 4 | 493 | 86 | |
| GenBank mRNA | AB297382.1 | - | 3 - 4 | 316 | 86 | |
| AB305045.1 | - | 3 - 4 | 316 | 86 | ||
| AK033388.1 | - | 3 - 4 | 435 | 86 | ||
| AK137755.1 | - | 3 - 4 | 425 | 86 | ||
| AK146984.1 | - | 3 - 4 | 400 | 86 | ||
| BC023845.1 | - | 3 - 4 | 497 | 5346000 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mboat1
Genomic Map